Learn More
Selleck Chemical LLC Malonic acid S3029-100mg
SDP
Catalog No. 503110735
Encompass_Preferred
Malonic acid (MOA) one of the major dicarboxylic acids (DCAs) in aerosols has been identified experimentally and computationally to be a strong acid Malonic acid (MOA) acts as a mediate bridge for the formation of pure SA-A-based clusters
Catalog No. 50-311-0735 Supplier Selleck Chemical LLC Supplier No. S3029100MG
Malonic acid (MOA) one of the major dicarboxylic acids (DCAs) in aerosols has been identified experimentally and computationally to be a strong acid Malonic acid (MOA) acts as a mediate bridge for the formation of pure SA-A-based clusters