Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Malonic acid, 99%, MP Biomedicals™

Catalog No. ICN10223880 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
250 g
500 g
1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10223880 100 g
ICN10223883 250 g
ICN10223890 500 g
ICN10223891 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN10223880 Supplier MP Biomedicals, Inc Supplier No. 0210223880
Add to Cart
Add to Cart

Chemical Identifiers

CAS 141-82-2
Molecular Formula C3H4O4
Molecular Weight (g/mol) 104.061
InChI Key OFOBLEOULBTSOW-UHFFFAOYSA-N
Synonym malonic acid, dicarboxymethane, carboxyacetic acid, methanedicarboxylic acid, kyselina malonova, usaf ek-695, dicarboxylate, dicarboxylic acid, kyselina malonova czech, propanediolic acid
PubChem CID 867
ChEBI CHEBI:30794
IUPAC Name propanedioic acid
SMILES C(C(=O)O)C(=O)O

Specifications

Color White
Quantity 100 g
Formula Weight 104.1
Percent Purity 0.99
Physical Form Powder
Chemical Name or Material Malonic acid
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.