Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Malononitrile

Catalog No. M1407100GA Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1407100GA 100 g
M1407500GA 500 g
M14075GA 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M1407100GA Supplier Sigma Aldrich Supplier No. M1407100GA
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Malononitrile

Specifications

CAS 109-77-3
Melting Point 30°C to 32°C (lit.)
Density 1.049 g/mL (at 25°C (literature))
Boiling Point 220°C (lit.)
Molecular Formula C3H2N2
Linear Formula CH2(CN)2
MDL Number MFCD00001883
Quantity 100 g
Synonym Dicyanomethane
Molecular Weight (g/mol) 66.06
Percent Purity ≥99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.