Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Malonyl chloride

Catalog No. M1601100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1601100G 100 g
M160125G 25 g
M16015G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M1601100G Supplier Sigma Aldrich Supplier No. M1601100G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Malonyl chloride can be used as a reactant for the synthesis of:
  • Alkaloids with tetracyclic cores such as cyclopiamide a and speradine e
  • 6-tetrathiafulvalene(ttf)-polymer by condensation polymerization with ttf-based dihydroxy monomer
  • N-doped tio2 films by molecular layer deposition
  • Alkyne-substituted 1,3-oxazines by reaction with arylpropynamides.it can also be used as a coupling agent in the synthesis of block copolymers by anionic polymerization

Specifications

CAS 1663-67-8
Density 1.449 g/mL (at 25°C (literature))
Boiling Point 53°C to 55°C (19 mmHg)
Molecular Formula C3H2Cl2O2
Refractive Index n20/D 1.465 (literature)
Linear Formula CH2(COCl)2
MDL Number MFCD00000735
Quantity 100 g
Synonym Malonyl dichloride
Molecular Weight (g/mol) 140.95
Percent Purity 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.