Learn More
Medchemexpress LLC Malonyl CoA lithium | 108347-84-8 | 98.0% | 1 MG
SDP
Catalog No. 5000351166
Encompass_Preferred
Malonyl CoA lithium is an inhibitor of carnitine palmitoyl transferase 1 (CPT1). High Malonyl CoA lithium concentrations suppress fatty acid oxidation, while low Malonyl CoA lithium concentrations are permissive for fat oxidation.
  • Inhibitor of carnitine palmitoyl transferase 1 (CPT1).
  • High concentrations suppress fatty acid oxidation.
  • Low concentrations are permissive for fat oxidation.
Catalog No. 50-003-51166 Supplier Medchemexpress LLC Supplier No. HY1364081MG
Malonyl CoA lithium is an inhibitor of carnitine palmitoyl transferase 1 (CPT1). High Malonyl CoA lithium concentrations suppress fatty acid oxidation, while low Malonyl CoA lithium concentrations are permissive for fat oxidation.
  • Inhibitor of carnitine palmitoyl transferase 1 (CPT1).
  • High concentrations suppress fatty acid oxidation.
  • Low concentrations are permissive for fat oxidation.