Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Malonyl CoA lithium | 108347-84-8 | 99.0% | C24H38N7O19P3S.xLi | 10MG
SDP

Catalog No. 5000258800
Encompass_Preferred

Malonyl CoA lithium is the lithium salt of malonyl coenzyme A used as a biochemical tool to modulate fatty acid metabolism. It functions as an inhibitor of carnitine palmitoyl transferase 1 (CPT1) and is provided for research applications with supporting documentation available.

  • Inhibits carnitine palmitoyl transferase 1 (CPT1).
  • Lithium salt of malonyl coenzyme A.
  • Supplied in research-scale quantities (1 mg-100 mg).
  • Purity 98.0-99.0% depending on batch.
  • Molecular formula C24H38N7O19P3S·xLi.
  • Data sheet and SDS available for handling and safety information.

Catalog No. 50-002-58800 Supplier Medchemexpress LLC Supplier No. HY13640810MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Malonyl CoA lithium is the lithium salt of malonyl coenzyme A used as a biochemical tool to modulate fatty acid metabolism. It functions as an inhibitor of carnitine palmitoyl transferase 1 (CPT1) and is provided for research applications with supporting documentation available.

  • Inhibits carnitine palmitoyl transferase 1 (CPT1).
  • Lithium salt of malonyl coenzyme A.
  • Supplied in research-scale quantities (1 mg-100 mg).
  • Purity 98.0-99.0% depending on batch.
  • Molecular formula C24H38N7O19P3S·xLi.
  • Data sheet and SDS available for handling and safety information.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.