Learn More
Medchemexpress LLC Malotilate | 59937-28-9 | 99.47% | C12H16O4S2 | 25 MG
SDP
Catalog No. 5000357831
Encompass_Preferred
Malotilate is an orally active hepatotropic and anti-fibrotic agent that selectively inhibits 5-lipoxygenase (5-LOX) with an IC50 of 4.7 μM. It prevents the development of hepatocytic injury in alcohol-pyrazole hepatitis by decreasing hepatic acetaldehyde levels and preventing transferrin retention in hepatocytes.
  • Selectively inhibits 5-lipoxygenase (5-LOX)
  • Prevents hepatocytic injury in alcohol-pyrazole hepatitis
  • Decreases hepatic acetaldehyde levels
  • Prevents retention of transferrin in hepatocytes
  • Reduces collagen synthesis and cell migration activity of fibroblasts in vitro
  • Prevents acute experimental liver injury induced by various hepatotoxic compounds
  • Normalizes reduced serum cholesterol levels in rats with carbon tetrachloride-induced liver damage
Catalog No. 50-003-57831 Supplier Medchemexpress LLC Supplier No. 25MGHYA0060
Malotilate is an orally active hepatotropic and anti-fibrotic agent that selectively inhibits 5-lipoxygenase (5-LOX) with an IC50 of 4.7 μM. It prevents the development of hepatocytic injury in alcohol-pyrazole hepatitis by decreasing hepatic acetaldehyde levels and preventing transferrin retention in hepatocytes.
  • Selectively inhibits 5-lipoxygenase (5-LOX)
  • Prevents hepatocytic injury in alcohol-pyrazole hepatitis
  • Decreases hepatic acetaldehyde levels
  • Prevents retention of transferrin in hepatocytes
  • Reduces collagen synthesis and cell migration activity of fibroblasts in vitro
  • Prevents acute experimental liver injury induced by various hepatotoxic compounds
  • Normalizes reduced serum cholesterol levels in rats with carbon tetrachloride-induced liver damage