Learn More
Medchemexpress LLC Malotilate | 59937-28-9 | 99.5% | C12H16O4S2 | 100 MG
SDP
Catalog No. 5000432320
Encompass_Preferred
Malotilate is an orally active hepatotropic agent and an anti-fibrotic substance. It selectively inhibits 5-lipoxygenase (5-LOX) with an IC50 of 4.7 μM. It prevents the development of hepatocytic injury in alcohol-pyrazole hepatitis by decreasing hepatic acetaldehyde levels and preventing the retention of transferrin in the hepatocytes. For research use only.
  • Reduces collagen synthesis and cell migration activity of fibroblasts in vitro
  • Stimulates 12- and 15-lipoxygenase pathways
  • Prevents acute experimental liver injury induced by various hepatotoxic compounds
  • Normalizes lowered serum cholesterol in rats with hypocholesterolemia
  • Normalizes inhibition of triglyceride secretion from livers in rats with CCl4-induced liver damage
Catalog No. 50-004-32320 Supplier Medchemexpress LLC Supplier No. HYA0060100MG
Malotilate is an orally active hepatotropic agent and an anti-fibrotic substance. It selectively inhibits 5-lipoxygenase (5-LOX) with an IC50 of 4.7 μM. It prevents the development of hepatocytic injury in alcohol-pyrazole hepatitis by decreasing hepatic acetaldehyde levels and preventing the retention of transferrin in the hepatocytes. For research use only.
  • Reduces collagen synthesis and cell migration activity of fibroblasts in vitro
  • Stimulates 12- and 15-lipoxygenase pathways
  • Prevents acute experimental liver injury induced by various hepatotoxic compounds
  • Normalizes lowered serum cholesterol in rats with hypocholesterolemia
  • Normalizes inhibition of triglyceride secretion from livers in rats with CCl4-induced liver damage