Learn More
Medchemexpress LLC Malotilate | 59937-28-9 | C12H16O4S2 | 50 MG
SDP
Catalog No. 5000432083
Encompass_Preferred
Malotilate is an orally active hepatotropic agent and an anti-fibrotic substance. It selectively inhibits 5-lipoxygenase (5-LOX) with an IC50 of 4.7 μM. It helps prevent the development of hepatocytic injury in alcohol-pyrazole hepatitis by reducing hepatic acetaldehyde levels and preventing the retention of transferrin in hepatocytes.
  • Selectively inhibits 5-lipoxygenase (5-LOX) (IC50=4.7 μM).
  • Prevents the development of hepatocytic injury in alcohol-pyrazole hepatitis by decreasing hepatic acetaldehyde levels.
  • Prevents the retention of transferrin in the hepatocytes.
Catalog No. 50-004-32083 Supplier Medchemexpress LLC Supplier No. HYA006050MG
Malotilate is an orally active hepatotropic agent and an anti-fibrotic substance. It selectively inhibits 5-lipoxygenase (5-LOX) with an IC50 of 4.7 μM. It helps prevent the development of hepatocytic injury in alcohol-pyrazole hepatitis by reducing hepatic acetaldehyde levels and preventing the retention of transferrin in hepatocytes.
  • Selectively inhibits 5-lipoxygenase (5-LOX) (IC50=4.7 μM).
  • Prevents the development of hepatocytic injury in alcohol-pyrazole hepatitis by decreasing hepatic acetaldehyde levels.
  • Prevents the retention of transferrin in the hepatocytes.