Learn More
Selleck Chemical LLC Malotilate 10mg 59937-28-9 NKK105
SDP
Catalog No. 507600
Encompass_Preferred
Malotilate (NKK105) is a drug used for the treatment of liver cirrhosis. Malotilate is a liver protein metabolism improved compound, which selectively inhibit the 5-lipoxygenase. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-760-0 Supplier Selleck Chemical LLC Supplier No. S113710MG
Malotilate (NKK105) is a drug used for the treatment of liver cirrhosis. Malotilate is a liver protein metabolism improved compound, which selectively inhibit the 5-lipoxygenase. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.