Learn More
Enzo Life Sciences MALP 2 (50 µg)
SDP
Supplier:  Enzo Life Sciences ALX162027C050
Encompass_Preferred
Activator of the TLR2/TLR6 complex. Alternative Name: Macrophage activating lipopeptide 2, S-[2,3-bis(Palmityloxy)-(2R)-propyl-cysteinyl-GNNDESNISFKEK]. Formula: C99H167N19O30S. MW: 2135.2. Formulation: Liquid. Sterile solution in PBS containing 2.5% (v/v) 2-propanol, 25mM n-octyl-β-D-glucopyranoside and 1% (wt/v) human serum albumin. Long Term Storage: -80°C. Use/Stability: Stable for at least 2 years when stored at -20°C. Handling: Avoid accidental injection, extreme care should be taken with hypodermic syringes. Avoid inhalation and prevent this compound from entering the bloodstream. Avoid freeze/thaw cycles.
Catalog No. NC9311891