Learn More
Enzo Life Sciences MALP-2 (BULK) (500µg)
SDP
Catalog No. 502007841
Encompass_Preferred
MALP-2 was originally isolated from Mycoplasma fermentans. This MALP-2 corresponds to the originally isolated isomer, which expresses potent endotoxin-like activity and approaches in certain experimental systems the toxicity of LPS. For description of the stereochemistry of MALP-2. MALP-2 signalling, unlike that of LPS, is not transduced via TLR4, but is induced via TLR2 and TLR6 signalling. BULK size (500μg) of biologically active, pyrogen-free and sterile MALP-2 for in vitro and animal in vivo studies. Alternative name: Macrophage activating lipopeptide 2, S-[2,3-bis(Palmityloxy)-(2R)-propyl-cysteinyl-GNNDESNISFKEK]. Formula: C99H167N19O30S. MW: 2135.2. Formulation: Liquid. Pyrogen-free, sterile solution in 33% (v/v) 2-propanol/water. Handling: Avoid accidental injection, extreme care should be taken with hypodermic syringes. Avoid inhalation and prevent this compound from entering the bloodstream. Avoid freeze/thaw cycles.
Catalog No. 50-200-7841 Supplier Enzo Life Sciences Supplier No. ALX162034C500
MALP-2 was originally isolated from Mycoplasma fermentans. This MALP-2 corresponds to the originally isolated isomer, which expresses potent endotoxin-like activity and approaches in certain experimental systems the toxicity of LPS. For description of the stereochemistry of MALP-2. MALP-2 signalling, unlike that of LPS, is not transduced via TLR4, but is induced via TLR2 and TLR6 signalling. BULK size (500μg) of biologically active, pyrogen-free and sterile MALP-2 for in vitro and animal in vivo studies. Alternative name: Macrophage activating lipopeptide 2, S-[2,3-bis(Palmityloxy)-(2R)-propyl-cysteinyl-GNNDESNISFKEK]. Formula: C99H167N19O30S. MW: 2135.2. Formulation: Liquid. Pyrogen-free, sterile solution in 33% (v/v) 2-propanol/water. Handling: Avoid accidental injection, extreme care should be taken with hypodermic syringes. Avoid inhalation and prevent this compound from entering the bloodstream. Avoid freeze/thaw cycles.