Learn More
NEOGEN Malt Extract, 10 kg
SDP
Catalog No. 502015740
Encompass_Preferred
Malt Extract is a dehydrated extract of malt for use in preparing microbiological culture media in a laboratory setting. Malt Extract is not intended for use in the diagnosis of disease or other conditions in humans.Malt Extract is a clarified, water-soluble extract of malted barley. Malt Extract is a useful ingredient of culture media designed for the propagation of yeasts and molds. This ingredient is suitable for yeasts and molds because it contains a high concentration of carbohydrates, particularly maltose. The approximate percentage of reducing sugars in Malt Extract is 60 – 63%.The high carbohydrate content makes malt extract very sensitive to overheating which results in darkening of the medium it is used in. Malt Extract is generally employed in culture media at concentrations between 10 to 100 grams per liter.For more information visit https://www.neogen.com/solutions/Microbiology/malt-extract
Catalog No. 50-201-5740 Supplier NEOGEN Supplier No. 700003508
Malt Extract is a dehydrated extract of malt for use in preparing microbiological culture media in a laboratory setting. Malt Extract is not intended for use in the diagnosis of disease or other conditions in humans.Malt Extract is a clarified, water-soluble extract of malted barley. Malt Extract is a useful ingredient of culture media designed for the propagation of yeasts and molds. This ingredient is suitable for yeasts and molds because it contains a high concentration of carbohydrates, particularly maltose. The approximate percentage of reducing sugars in Malt Extract is 60 – 63%.The high carbohydrate content makes malt extract very sensitive to overheating which results in darkening of the medium it is used in. Malt Extract is generally employed in culture media at concentrations between 10 to 100 grams per liter.For more information visit https://www.neogen.com/solutions/Microbiology/malt-extract