Learn More
Alkali Scientific Malt Extract, 500 G
SDP
Catalog No. 5000661714
Encompass_Preferred
Malt Extract (500 g) is a versatile nutrient medium used to support the growth of fungi, particularly yeasts and molds. This extract is rich in sugars and provides the necessary nutrients to promote fungal growth in laboratory cultures. Malt Extract is widely used in the microbiological industry for cultivating microorganisms in fermentation processes and for conducting research on fungal metabolism and growth. It is also commonly used in the production of selective media for fungal isolation. This medium is essential for microbiology, industrial applications, and food microbiology where fungal growth plays a critical role.
Catalog No. 50-006-61714 Supplier Alkali Scientific Supplier No. XTR003CP
Malt Extract (500 g) is a versatile nutrient medium used to support the growth of fungi, particularly yeasts and molds. This extract is rich in sugars and provides the necessary nutrients to promote fungal growth in laboratory cultures. Malt Extract is widely used in the microbiological industry for cultivating microorganisms in fermentation processes and for conducting research on fungal metabolism and growth. It is also commonly used in the production of selective media for fungal isolation. This medium is essential for microbiology, industrial applications, and food microbiology where fungal growth plays a critical role.