Learn More
Research Products International Corp Malt Extract, 500 Grams
SDP
Catalog No. 50997923
Encompass_Preferred
Malt Extract is used for the cultivation and maintenance of yeasts and molds.
Catalog No. 50-997-923 Supplier Research Products International Corp Supplier No. M41600500.0
Malt Extract is used for the cultivation and maintenance of yeasts and molds.