Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Research Products International Corp Malt Extract Agar 5 KG
SDP

Catalog No. 502230466
Encompass_Preferred

Malt Extract Agar is used for the isolation, cultivation and maintenance of yeasts and molds. Formulation is similar to Malt Extract Broth with the addition of agar and glycerol. This broth medium was recommended by Thom and Church for the cultivation of fungi. Its low acidity inhibits the growth of most bacteria. The high carbohydrate content enhances the rapid growth of fungi and yeasts.

Dissolve 33.6 grams in 1 liter of distilled water. Sterilize by autoclaving for 15 minutes.

Ingredients:

Maltose: 12.75 g/L
Dextrin: 2.75 g/L
Glycerol: 2.35 g/L
Peptone: 0.78 g/L
Agar: 15.0 g/L
Final pH (25°C): 4.6 ± 0.2

Catalog No. 50-223-0466 Supplier Research Products International Corp Supplier No. M215005000.0
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Malt Extract Agar is used for the isolation, cultivation and maintenance of yeasts and molds. Formulation is similar to Malt Extract Broth with the addition of agar and glycerol. This broth medium was recommended by Thom and Church for the cultivation of fungi. Its low acidity inhibits the growth of most bacteria. The high carbohydrate content enhances the rapid growth of fungi and yeasts.

Dissolve 33.6 grams in 1 liter of distilled water. Sterilize by autoclaving for 15 minutes.

Ingredients:

Maltose: 12.75 g/L
Dextrin: 2.75 g/L
Glycerol: 2.35 g/L
Peptone: 0.78 g/L
Agar: 15.0 g/L
Final pH (25°C): 4.6 ± 0.2

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.