Learn More
NEOGEN Malt Extract Agar, 500 g
SDP
Catalog No. 502015426
Encompass_Preferred
Malt Extract Agar is used for the cultivation of fungi and is not intended for use in the diagnosis of disease or other conditions in humans. An acidic medium which will support the growth of most yeasts and molds whilst inhibiting most bacteria. It was first described by Thom and Church in 1926 in a study of Aspergillus spp. claiming the high carbohydrate content ensured rapid growth. Selectivity can be increased by further lowering the pH with the addition, after sterilization, of Lactic Acid (X037). It should be noted that excess heating of this medium together with its low pH can easily result in hydrolysis of the agar gel producing soft plates.  Formula:Malt Extract - 30.0 g/LPeptone - 5.0 g/LAgar - 15.0 g/LFor more information visit https://www.neogen.com/solutions/Microbiology/malt-extract-agar
Catalog No. 50-201-5426 Supplier NEOGEN Supplier No. 700004486
Malt Extract Agar is used for the cultivation of fungi and is not intended for use in the diagnosis of disease or other conditions in humans. An acidic medium which will support the growth of most yeasts and molds whilst inhibiting most bacteria. It was first described by Thom and Church in 1926 in a study of Aspergillus spp. claiming the high carbohydrate content ensured rapid growth. Selectivity can be increased by further lowering the pH with the addition, after sterilization, of Lactic Acid (X037). It should be noted that excess heating of this medium together with its low pH can easily result in hydrolysis of the agar gel producing soft plates.  Formula:Malt Extract - 30.0 g/LPeptone - 5.0 g/LAgar - 15.0 g/LFor more information visit https://www.neogen.com/solutions/Microbiology/malt-extract-agar