Learn More
Bioworld Malt Extract Agar, 500 g
Catalog No. 50254470
Encompass_Preferred
Malt Extract AgarUsed for the isolation, enumeration, and cultivation of yeasts, molds and other aciduric microorganismspH: 4.7Dehydrated Appearance: off-white, homogeneous powderPrepared Appearance: light amber agar Soluble in boiling waterComposition (Per Liter):Malt Extract: 30gPeptone (meat peptone): 5gAgar: 15gpH 7.2 ± 0.2 @ 37°C
Catalog No. 50-254-470 Supplier Bioworld Supplier No. 306260011
Malt Extract AgarUsed for the isolation, enumeration, and cultivation of yeasts, molds and other aciduric microorganismspH: 4.7Dehydrated Appearance: off-white, homogeneous powderPrepared Appearance: light amber agar Soluble in boiling waterComposition (Per Liter):Malt Extract: 30gPeptone (meat peptone): 5gAgar: 15gpH 7.2 ± 0.2 @ 37°C