Learn More
MOLTOX Molecular Toxicology Inc Malt Extract Agar, 20 mL/plate, 20 plates/sleeve, Store in Refrigerator
SDP
Catalog No. 50948676
Encompass_Preferred
Malt Extract Agar is most commonly used for the propagation of yeasts and molds. Other uses include isolation, cultivation and the enumeration of yeasts and molds. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.
Catalog No. 50-948-676 Supplier MOLTOX Molecular Toxicology Inc Supplier No. 2140S60
Malt Extract Agar is most commonly used for the propagation of yeasts and molds. Other uses include isolation, cultivation and the enumeration of yeasts and molds. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.