Learn More
Teknova Malt Extract Agar Plates. 100mm, 10 Plates per Sleeve, Sterile. 4 pack.
SDP
Catalog No. 502387673
Encompass
Malt Extract Media is a liquid broth or agar commonly used to culture bacteria. It is also particularly useful for isolating and identifying different species of yeasts and molds.
Catalog No. 50-238-7673 Supplier Teknova Supplier No. 2M00434PK
Malt Extract Media is a liquid broth or agar commonly used to culture bacteria. It is also particularly useful for isolating and identifying different species of yeasts and molds.