Learn More
Teknova Malt Extract Agar Plates. 100mm Plates, 20 Plates per Sleeve, Sterile.
SDP
Catalog No. 502388171
Encompass
Malt Extract Media is a liquid broth or agar commonly used to culture bacteria. It is also particularly useful for isolating and identifying different species of yeasts and molds.
Catalog No. 50-238-8171 Supplier Teknova Supplier No. M0043
Malt Extract Media is a liquid broth or agar commonly used to culture bacteria. It is also particularly useful for isolating and identifying different species of yeasts and molds.