Learn More
Bioworld Malt Extract Broth with Dextrose, 500 g
Catalog No. 50254331
Encompass_Preferred
Malt Extract Broth Used for the cultivation and maintenance of yeasts and molds pH: 4.7 Working Concentration: 15 g/L Dehydrated Appearance: light beige to beige, homogeneous powder Prepared Appearance: light amber broth Soluble in water Contains: Malt Extract Base Maltose Dextrose Yeast Extract Usually ships within 24 hours. This product is sold for laboratory research use only.
Catalog No. 50-254-331 Supplier Bioworld Supplier No. 306200501
Malt Extract Broth Used for the cultivation and maintenance of yeasts and molds pH: 4.7 Working Concentration: 15 g/L Dehydrated Appearance: light beige to beige, homogeneous powder Prepared Appearance: light amber broth Soluble in water Contains: Malt Extract Base Maltose Dextrose Yeast Extract Usually ships within 24 hours. This product is sold for laboratory research use only.