Learn More
Bioworld Malt Extract Broth with Dextrose, 1 kg
Catalog No. 50254332
Encompass_Preferred
Malt Extract BrothUsed for the cultivation and maintenance of yeasts and moldspH: 4.7Working Concentration: 15 g/LDehydrated Appearance: light beige to beige, homogeneous powderPrepared Appearance: light amber brothSoluble in waterContains: Malt Extract Base Maltose Dextrose Yeast Extract Usually ships within 24 hours. This product is sold for laboratory research use only.
Catalog No. 50-254-332 Supplier Bioworld Supplier No. 306200502
Malt Extract BrothUsed for the cultivation and maintenance of yeasts and moldspH: 4.7Working Concentration: 15 g/LDehydrated Appearance: light beige to beige, homogeneous powderPrepared Appearance: light amber brothSoluble in waterContains: Malt Extract Base Maltose Dextrose Yeast Extract Usually ships within 24 hours. This product is sold for laboratory research use only.