Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Research Products International Corp Malt Extract Broth, 500 Grams
SDP

Catalog No. 50488803
Encompass_Preferred

Medium was recommended by Thom and Church for the cultivation of fungi. Its low acidity inhibits the growth of most bacteria. The high carbohydrate content enhances the rapid growth of fungi and yeasts.

Dissolve 15.0 g in 1 liter of distilled water. Sterilize by autoclaving for 15 minutes.

Ingredients:

Malt Extract Base: 6.0 g/L
Dextrose: 6.0 g/L

Maltose: 1.8g/L
Yeast: 1.2 g/L

Catalog No. 50-488-803 Supplier Research Products International Corp Supplier No. M22500500.0
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Medium was recommended by Thom and Church for the cultivation of fungi. Its low acidity inhibits the growth of most bacteria. The high carbohydrate content enhances the rapid growth of fungi and yeasts.

Dissolve 15.0 g in 1 liter of distilled water. Sterilize by autoclaving for 15 minutes.

Ingredients:

Malt Extract Base: 6.0 g/L
Dextrose: 6.0 g/L

Maltose: 1.8g/L
Yeast: 1.2 g/L

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.