Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Malt Extract, MP Biomedicals

Catalog No. ICN15531590 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15531590 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15531590 Supplier MP Biomedicals, Inc Supplier No. 0215531590
Add to Cart
Add to Cart

From starch digestion. A source of carbohydrates

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8002-48-0

Specifications

Quantity 500 g
Chemical Name or Material Malt Extract
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.