Learn More
enQuire Bioreagents MALT1 Anti-Human Monoclonal [Clone MT1/410], Size: 100ug
Supplier:  enQuire Bioreagents 10892MSM1P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, WB. Conjugate/Tag: Unconjugated. Clone: MT1/410. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: Jurkat, Daudi or HeLa cells. Tonsil or lymphoma. Cellular Staining: Cytoplasmic. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Human MALT1 recombinant fragment (aa701-808). Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 93kDa.Chromosome: 18q21.32. Gene Symbol: MALT1. Gene ID: 10892. Ensemble: ENSG00000172175. Uniprot: NP_776216.
Catalog No. 50-188-4596