Learn More
Enzo Life Sciences Z-VRPR-FMK (1 mg)
SDP
Catalog No. NC9073082
Encompass_Preferred
MALT1 inhibitor. The proteolytic activity of the paracaspase MALT1 is key to NF-κB and T cell activation. Z-VRPR-FMK is a selective MALT1 inhibitor of such proteolytic activity. Alternative Name: Z-Val-Arg-Pro-DL-Arg-FMK, MALT1 inhibitor. Formula: C31H49FN10O6. MW: 676.8. 114. Purity: ≥90% (HPLC). Sequence: Z-Val-Arg-Pro-DL-Arg-fluoromethylketone . TFA. Appearance: White to slightly yellow powder. Solubility: Soluble in water (1mg/ml). Long Term Storage: -20°C. Handling: Hygroscopic. Avoid freeze/thaw cycles.
Catalog No. NC9073082 Supplier Enzo Life Sciences Supplier No. ALX260166M001
MALT1 inhibitor. The proteolytic activity of the paracaspase MALT1 is key to NF-κB and T cell activation. Z-VRPR-FMK is a selective MALT1 inhibitor of such proteolytic activity. Alternative Name: Z-Val-Arg-Pro-DL-Arg-FMK, MALT1 inhibitor. Formula: C31H49FN10O6. MW: 676.8. 114. Purity: ≥90% (HPLC). Sequence: Z-Val-Arg-Pro-DL-Arg-fluoromethylketone . TFA. Appearance: White to slightly yellow powder. Solubility: Soluble in water (1mg/ml). Long Term Storage: -20°C. Handling: Hygroscopic. Avoid freeze/thaw cycles.