Learn More
Abnova™ MALT1 Split CISH Probe
Catalog No. 89335549 Shop All Abnova Corporation Products
Change view
Click to view available options
For Use With (Application):
CISH-Ce
Catalog No. For Use With (Application)
89-335-549 CISH-Ce
1 options
Catalog No. 89-335-549 Supplier Abnova™ Supplier No. CS0012
Add to Cart
Add to Cart
MALT1 Split CISH Probe is designed for the qualitative detection of human MALT1 gene at 18q21.32 in formalin-fixed, paraffin-embedded specimens by chromogenic in situ hybridization (CISH).
The probe is intended to be used in combination with the CISH Implementation Kit 2, which provides necessary reagents for specimen pretreatment and post-hybridization processing.
Hybridization signals of digoxigenin-labeled polynucleotides appear dark green distinct dot-shaped (proximal to the MALT1 breakpoint region), and dinitrophenyl-labeled polynucleotides appear bright red distinct dot-shaped (distal to the MALT1 breakpoint region).
Normal situation: In interphases of normal cells or cells without a translocation involving the MALT1 gene region, two red/green fusion signals appear.
Aberrant situation: One MALT1 gene region affected by a translocation is indicated by one separate green signal and one separate red signal. Other signal distribution may be observed in some abnormal samples which might result in a different signal pattern than described above, indicating variant rearrangements.
Unexpected signal patterns should be further investigated.
Specifications
Content And Storage The product is ready-to-use. No reconstitution, mixing, or dilution is required. Bring probe to room temperature (18 to 25°C) and mix briefly before use.
For Use With (Application) CISH-Ce