Learn More
Medchemexpress LLC HY-B2122 500mg Medchemexpress, Maltitol CAS:585-88-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB2122500MG
Encompass_Preferred
Medchemexpress, HY-B2122 500mg Maltitol CAS:585-88-6 Maltitol is a sugar alcohol used as a sugar substitute. It has 75-90% of the sweetness of sucrose (table sugar) and nearly identical properties. Maltitol may also be used as a plasticizer in gelatin capsules, as an emollient, and as a humectant. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2724