Learn More
Medchemexpress LLC Maltitol | 585-88-6 | 98.0% | 1 ML
SDP
Catalog No. 5000411654
Encompass_Preferred
Maltitol is an orally active polyol and sugar alcohol sweetener derived from the hydrogenation of maltose. It can serve as a substitute for sucrose and is notable for its minor impact on blood sugar levels and potential prebiotic properties.
  • Can replace sucrose
  • Minor impact on blood sugar
  • Potential prebiotic effects
  • Prevents nonalcoholic fatty liver disease (NAFLD) in mouse models
  • Not genotoxic or cytotoxic in rats
  • Does not induce chromosomal aberrations or reduce mitotic index
Catalog No. 50-004-11654 Supplier Medchemexpress LLC Supplier No. HYB212210MM/1ML
Maltitol is an orally active polyol and sugar alcohol sweetener derived from the hydrogenation of maltose. It can serve as a substitute for sucrose and is notable for its minor impact on blood sugar levels and potential prebiotic properties.
  • Can replace sucrose
  • Minor impact on blood sugar
  • Potential prebiotic effects
  • Prevents nonalcoholic fatty liver disease (NAFLD) in mouse models
  • Not genotoxic or cytotoxic in rats
  • Does not induce chromosomal aberrations or reduce mitotic index