Learn More
Medchemexpress LLC Maltitol (Standard) | 585-88-6 | 96.15% | 50 MG
SDP
Catalog No. 5000411671
Encompass_Preferred
Maltitol (Standard) is the analytical standard of Maltitol (HY-B2122) and is intended for research and analytical applications. It is an orally active polyol and sugar alcohol sweetener produced by the hydrogenation of maltose. It can replace sucrose and has characteristics of having a minor impact on blood sugar and potential prebiotic effects.
  • Used for qualitative, quantitative, and methodological research experiments.
  • Suitable for HPLC, GC, and MS applications.
  • Has a minor impact on blood sugar.
  • Offers potential prebiotic effects.
Catalog No. 50-004-11671 Supplier Medchemexpress LLC Supplier No. HYB2122R50MG
Maltitol (Standard) is the analytical standard of Maltitol (HY-B2122) and is intended for research and analytical applications. It is an orally active polyol and sugar alcohol sweetener produced by the hydrogenation of maltose. It can replace sucrose and has characteristics of having a minor impact on blood sugar and potential prebiotic effects.
  • Used for qualitative, quantitative, and methodological research experiments.
  • Suitable for HPLC, GC, and MS applications.
  • Has a minor impact on blood sugar.
  • Offers potential prebiotic effects.