Learn More
Medchemexpress LLC Maltitol (standard) | 585-88-6 | 96.15% | 500 MG
SDP
Catalog No. 5000411714
Encompass_Preferred
Maltitol is an orally active polyol and sugar alcohol sweetener produced by the hydrogenation of maltose. It can replace sucrose and has characteristics of having a minor impact on blood sugar and potential prebiotic effects. This analytical standard is intended for research and analytical applications.
  • Analytical standard for research and analytical applications
  • Orally active polyol and sugar alcohol sweetener
  • Can replace sucrose
  • Minor impact on blood sugar
  • Potential prebiotic effects
  • Commonly used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS
Catalog No. 50-004-11714 Supplier Medchemexpress LLC Supplier No. HYB2122R500MG
Maltitol is an orally active polyol and sugar alcohol sweetener produced by the hydrogenation of maltose. It can replace sucrose and has characteristics of having a minor impact on blood sugar and potential prebiotic effects. This analytical standard is intended for research and analytical applications.
  • Analytical standard for research and analytical applications
  • Orally active polyol and sugar alcohol sweetener
  • Can replace sucrose
  • Minor impact on blood sugar
  • Potential prebiotic effects
  • Commonly used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS