Learn More
Medchemexpress LLC Maltitol (Standard) | 585-88-6 | 96.2% | 100 MG
SDP
Catalog No. 5000411713
Encompass_Preferred
Maltitol (Standard) is the analytical standard of Maltitol (HY-B2122). It is an orally active polyol and sugar alcohol sweetener produced by the hydrogenation of maltose. It can replace sucrose and has characteristics of having a minor impact on blood sugar and potential prebiotic effects.
  • Intended for research and analytical applications.
  • Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Has a minor impact on blood sugar.
  • Has potential prebiotic effects.
  • Can replace sucrose.
Catalog No. 50-004-11713 Supplier Medchemexpress LLC Supplier No. HYB2122R100MG
Maltitol (Standard) is the analytical standard of Maltitol (HY-B2122). It is an orally active polyol and sugar alcohol sweetener produced by the hydrogenation of maltose. It can replace sucrose and has characteristics of having a minor impact on blood sugar and potential prebiotic effects.
  • Intended for research and analytical applications.
  • Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Has a minor impact on blood sugar.
  • Has potential prebiotic effects.
  • Can replace sucrose.