Learn More
Medchemexpress LLC Maltitol (Standard) | 585-88-6 | 96.2% | 25 MG
SDP
Catalog No. 5000413538
Encompass_Preferred
Maltitol (Standard) serves as the analytical standard for Maltitol (HY-B2122) and is designed for research and analytical applications. It is an orally active polyol and sugar alcohol sweetener, synthesized through the hydrogenation of maltose. This product can be used as a substitute for sucrose and is recognized for its minimal effect on blood sugar levels and potential prebiotic qualities.
  • Analytical standard for Maltitol.
  • Intended for research and analytical applications.
  • Orally active polyol and sugar alcohol sweetener.
  • Can replace sucrose.
  • Has a minor impact on blood sugar.
  • Possesses potential prebiotic effects.
  • Commonly utilized in qualitative, quantitative, and methodological research experiments involving techniques such as HPLC, GC, and MS.
Catalog No. 50-004-13538 Supplier Medchemexpress LLC Supplier No. 25MGHYB2122R
Maltitol (Standard) serves as the analytical standard for Maltitol (HY-B2122) and is designed for research and analytical applications. It is an orally active polyol and sugar alcohol sweetener, synthesized through the hydrogenation of maltose. This product can be used as a substitute for sucrose and is recognized for its minimal effect on blood sugar levels and potential prebiotic qualities.
  • Analytical standard for Maltitol.
  • Intended for research and analytical applications.
  • Orally active polyol and sugar alcohol sweetener.
  • Can replace sucrose.
  • Has a minor impact on blood sugar.
  • Possesses potential prebiotic effects.
  • Commonly utilized in qualitative, quantitative, and methodological research experiments involving techniques such as HPLC, GC, and MS.