Learn More
Medchemexpress LLC Maltitol (Standard) | 585-88-6 | 98.0% | 10 G
SDP
Catalog No. 5000410341
Encompass_Preferred
Maltitol is an orally active polyol and sugar alcohol sweetener produced by the hydrogenation of maltose. It can replace sucrose and has characteristics of having a minor impact on blood sugar and potential prebiotic effects.
  • Can replace sucrose.
  • Has a minor impact on blood sugar.
  • Potential prebiotic effects.
Catalog No. 50-004-10341 Supplier Medchemexpress LLC Supplier No. HYB212210G
Maltitol is an orally active polyol and sugar alcohol sweetener produced by the hydrogenation of maltose. It can replace sucrose and has characteristics of having a minor impact on blood sugar and potential prebiotic effects.
  • Can replace sucrose.
  • Has a minor impact on blood sugar.
  • Potential prebiotic effects.