Learn More
Medchemexpress LLC Maltodextrin | 9050-36-6 | MFCD00146679 | C6nH(10n+2)O(5n+1) | 500 G
SDP
Catalog No. 5000281244
Encompass_Preferred
Maltodextrin (dextrose equivalent 16.5-19.5) is a polysaccharide-grade excipient provided as a dry powder for pharmaceutical formulation and delivery research. It serves as a stabilizer and encapsulation matrix for proteins and small molecules, improving solubility and processing characteristics while showing low cytotoxicity in typical test concentrations.
  • Provides stabilizing excipient functionality in formulations.
  • Enhances solubility and processability of active ingredients.
  • Supports protein encapsulation with ~70% encapsulation efficiency and up to 20% loading.
  • Exhibits low cytotoxicity at concentrations up to 5 mg/mL.
  • DE value of 16.5-19.5 indicates the degree of hydrolysis.
  • Available as a 500 G pack for laboratory use.
Catalog No. 50-002-81244 Supplier Medchemexpress LLC Supplier No. HYW250795500G
Maltodextrin (dextrose equivalent 16.5-19.5) is a polysaccharide-grade excipient provided as a dry powder for pharmaceutical formulation and delivery research. It serves as a stabilizer and encapsulation matrix for proteins and small molecules, improving solubility and processing characteristics while showing low cytotoxicity in typical test concentrations.
  • Provides stabilizing excipient functionality in formulations.
  • Enhances solubility and processability of active ingredients.
  • Supports protein encapsulation with ~70% encapsulation efficiency and up to 20% loading.
  • Exhibits low cytotoxicity at concentrations up to 5 mg/mL.
  • DE value of 16.5-19.5 indicates the degree of hydrolysis.
  • Available as a 500 G pack for laboratory use.