Learn More
Medchemexpress LLC Maltodextrin | 9050-36-6 | MFCD00146679 | Variable (no fixed molecular weight; polymeric - represented as (C6H10O5)n·xH2O) | (C6H10O5)n·xH2O | 100 G
SDP
Catalog No. 5000280510
Encompass_Preferred
Maltodextrin, dextrose equivalent 16.5-19.5 is a water-soluble polysaccharide used as an excipient and formulation aid in research applications. It provides intermediate hydrolysis (DE 16.5-19.5) and is supplied as a white to off-white powder for laboratory use.
  • Water-soluble excipient for stabilization and encapsulation.
  • Intermediate dextrose equivalent for balanced solubility and viscosity.
  • Forms nanoparticles with narrow size distribution for delivery studies.
  • Non-toxic at tested concentrations in cell assays.
  • Available in common laboratory pack sizes for research use.
Catalog No. 50-002-80510 Supplier Medchemexpress LLC Supplier No. HYW250795100G
Maltodextrin, dextrose equivalent 16.5-19.5 is a water-soluble polysaccharide used as an excipient and formulation aid in research applications. It provides intermediate hydrolysis (DE 16.5-19.5) and is supplied as a white to off-white powder for laboratory use.
  • Water-soluble excipient for stabilization and encapsulation.
  • Intermediate dextrose equivalent for balanced solubility and viscosity.
  • Forms nanoparticles with narrow size distribution for delivery studies.
  • Non-toxic at tested concentrations in cell assays.
  • Available in common laboratory pack sizes for research use.