Learn More
Cayman Chemical Maltoheptaose, Purity: ≥80%, CAS: 34620-78-5, 50 mg
SDP
Catalog No. 502984078
Encompass_Preferred
A maltooligosaccharide consisting of seven glucose units
Catalog No. 50-298-4078 Supplier Cayman Chemical Supplier No. 2075850 MG
A maltooligosaccharide consisting of seven glucose units