Learn More
Medchemexpress LLC Maltol | 118-71-8 | MFCD00006578 | >=99.0% | 126.11 | 100 G
SDP
Catalog No. 5000454860
Encompass_Preferred
Maltol is an aromatic compound with antioxidant properties. It enhances neural function by mitigating oxidative stress and cell apoptosis. Maltol inhibits oxidative damage in nerve cells and helps prevent diabetic peripheral neuropathy (DPN). It is also widely used as a safe flavoring agent and food preservative. Additionally, Maltol acts as a metal ion chelator, useful in catalysis, cosmetics, and medicine.
  • Antioxidant agent
  • Enhances neural function by mitigating oxidative stress and cell apoptosis
  • Inhibitor of oxidative damage in nerve cells
  • Effective in preventing diabetic peripheral neuropathy (DPN)
  • Used as a flavoring agent and food preservative
  • Metal ion chelator
Catalog No. 50-004-54860 Supplier Medchemexpress LLC Supplier No. HYW012788100G
Maltol is an aromatic compound with antioxidant properties. It enhances neural function by mitigating oxidative stress and cell apoptosis. Maltol inhibits oxidative damage in nerve cells and helps prevent diabetic peripheral neuropathy (DPN). It is also widely used as a safe flavoring agent and food preservative. Additionally, Maltol acts as a metal ion chelator, useful in catalysis, cosmetics, and medicine.
  • Antioxidant agent
  • Enhances neural function by mitigating oxidative stress and cell apoptosis
  • Inhibitor of oxidative damage in nerve cells
  • Effective in preventing diabetic peripheral neuropathy (DPN)
  • Used as a flavoring agent and food preservative
  • Metal ion chelator