Learn More
Carbosynth LLC. Maltopentaose, 34620-76-3, MFCD00070166, 25 mg
SDP
Supplier:  Carbosynth LLC. OM0687225MG
Encompass_Preferred
Chemical Formula: C30H52O26, MW: 828.72, Synonyms: 4-O-[4-O-[4-O-(4-O-d-D-Glucopyranosyl-a-D-glucopyranosyl)-a-D-glucopyranosyl]-a-D-glucopyranosyl]-a-D-glucopyranose; Amylopentaose. Derived from starch by hydrolysis and chromatography.
Catalog No. NC1106490