Learn More
Cayman Chemical Maltopentaose 50mg
SDP
Catalog No. 502937797
Encompass_Preferred
An oligosaccharide comprised of five α-1,4-linked glucose molecules; increases the α-amylase synthesis rate in B. stearothermophilus at 1 mM; has been used as a substrate for PPA to study various inhibitors of PPA
Catalog No. 50-293-7797 Supplier Cayman Chemical Supplier No. 2845650 MG
An oligosaccharide comprised of five α-1,4-linked glucose molecules; increases the α-amylase synthesis rate in B. stearothermophilus at 1 mM; has been used as a substrate for PPA to study various inhibitors of PPA