Learn More
Medchemexpress LLC HY-N1495 5mg Medchemexpress, Maltopentaose CAS:34620-76-3 Purity:>98%
SDP
Catalog No. 501933844
Encompass_Preferred
Medchemexpress, HY-N1495 5mg Maltopentaose CAS:34620-76-3 Maltopentaose is the shortest chain oligosaccharide that can be classified as maltodextrin and is also used in a study to investigate glycation and phosphorylation of α-lactalbumin. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3844 Supplier Medchemexpress LLC Supplier No. HYN14955MG
Medchemexpress, HY-N1495 5mg Maltopentaose CAS:34620-76-3 Maltopentaose is the shortest chain oligosaccharide that can be classified as maltodextrin and is also used in a study to investigate glycation and phosphorylation of α-lactalbumin. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.