Learn More
Medchemexpress LLC Maltose-13C12 monohydrate | 372.22 | 5 MG
SDP
Catalog No. 5000414638
Encompass_Preferred
Maltose monohydrate-13C12 is the 13C labeled isotope of Maltose monohydrate. This compound can be used as a tracer or as an internal standard for quantitative analysis. It is suitable for use with analytical techniques such as NMR, GC-MS, or LC-MS.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis
  • Compatible with NMR
  • Compatible with GC-MS
  • Compatible with LC-MS
Catalog No. 50-004-14638 Supplier Medchemexpress LLC Supplier No. HYN2024AS15MG
Maltose monohydrate-13C12 is the 13C labeled isotope of Maltose monohydrate. This compound can be used as a tracer or as an internal standard for quantitative analysis. It is suitable for use with analytical techniques such as NMR, GC-MS, or LC-MS.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis
  • Compatible with NMR
  • Compatible with GC-MS
  • Compatible with LC-MS