Learn More
Medchemexpress LLC Maltose | 69-79-4 | 342.30 | 5 G
SDP
Catalog No. 5000415881
Encompass_Preferred
Maltose is a disaccharide formed from two units of glucose joined with an α(1→4) bond, a reducing sugar. Maltose monohydrate can be used as an energy source for bacteria. It is the two-unit member of the amylose homologous series, which is the key structural motif of starch. When beta-amylase breaks down starch, it removes two glucose units at a time, producing maltose. In the isomer isomaltose, the two glucose molecules are joined with an α(1→6) bond.
  • For research use only.
Catalog No. 50-004-15881 Supplier Medchemexpress LLC Supplier No. HYN20245G
Maltose is a disaccharide formed from two units of glucose joined with an α(1→4) bond, a reducing sugar. Maltose monohydrate can be used as an energy source for bacteria. It is the two-unit member of the amylose homologous series, which is the key structural motif of starch. When beta-amylase breaks down starch, it removes two glucose units at a time, producing maltose. In the isomer isomaltose, the two glucose molecules are joined with an α(1→6) bond.
  • For research use only.