Learn More
Medchemexpress LLC Maltose | 69-79-4 | MFCD00135877 | 342.30 | 1 G
SDP
Catalog No. 5000416271
Encompass_Preferred
Maltose is a disaccharide composed of two glucose units linked by an α(1→4) bond, functioning as a reducing sugar. It serves as an energy source for bacteria and is recognized as an analytical standard for research and analytical applications.
  • Formed from two glucose units joined with an α(1→4) bond.
  • Functions as a reducing sugar.
  • Can be used as an energy source for bacteria.
  • Intended for research and analytical applications.
Catalog No. 50-004-16271 Supplier Medchemexpress LLC Supplier No. HYN20241G
Maltose is a disaccharide composed of two glucose units linked by an α(1→4) bond, functioning as a reducing sugar. It serves as an energy source for bacteria and is recognized as an analytical standard for research and analytical applications.
  • Formed from two glucose units joined with an α(1→4) bond.
  • Functions as a reducing sugar.
  • Can be used as an energy source for bacteria.
  • Intended for research and analytical applications.