Learn More
Abcam Maltose Binding Protein Affinity Resin - Amintra, 100ML
Catalog No. 502656164
Encompass_Preferred
Product overview MBP tagging of recombinant proteins can result in increased expression levels and higher solubility of the target protein, while allowing high purity recovery.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-265-6164 Supplier Abcam Supplier No. AB270538100ML
Product overview MBP tagging of recombinant proteins can result in increased expression levels and higher solubility of the target protein, while allowing high purity recovery.
The product is subject to the following: Abcam Restricted Use Statement