Learn More
Medchemexpress LLC Maltose monohydrate-13C12 | 6363-53-7 | 372.22 | 1 MG
SDP
Catalog No. 5000416094
Encompass_Preferred
Maltose-13C12 monohydrate is a 13C labeled isotope of Maltose monohydrate. It is for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes are incorporated into drug molecules for quantitation during drug development.
  • Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-16094 Supplier Medchemexpress LLC Supplier No. HYN2024AS11MG
Maltose-13C12 monohydrate is a 13C labeled isotope of Maltose monohydrate. It is for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes are incorporated into drug molecules for quantitation during drug development.
  • Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.