Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Maltose monohydrate-13C12 | 6363-53-7 | 99.5% | 372.22 g/mol | 13C12H24O12 | 10 MG
SDP

Catalog No. 5000295479
Encompass_Preferred

Maltose-13C12 monohydrate is the uniformly 13C-labeled isotopologue of maltose monohydrate provided for research applications. It is intended for use as an isotopic tracer and internal standard in metabolic flux analysis and mass spectrometry, enabling differentiation from endogenous unlabeled maltose.

  • Uniform 13C12 labeling of all 12 carbon atoms
  • Molecular weight 372.22 g/mol
  • Purity 99.5%
  • Supplied as a monohydrate solid suitable for analytical workflows
  • Available in small-mass vials for tracer and standard preparation

Catalog No. 50-002-95479 Supplier Medchemexpress LLC Supplier No. HYN2024AS110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Maltose-13C12 monohydrate is the uniformly 13C-labeled isotopologue of maltose monohydrate provided for research applications. It is intended for use as an isotopic tracer and internal standard in metabolic flux analysis and mass spectrometry, enabling differentiation from endogenous unlabeled maltose.

  • Uniform 13C12 labeling of all 12 carbon atoms
  • Molecular weight 372.22 g/mol
  • Purity 99.5%
  • Supplied as a monohydrate solid suitable for analytical workflows
  • Available in small-mass vials for tracer and standard preparation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.