Learn More
Selleck Chemical LLC Maltose monohydrate S4496-100mg
SDP
Catalog No. 503112197
Encompass_Preferred
Maltose (Maltobiose Malt Sugar Beta-Maltose D-Maltose) monohydrate an non-essential nutrient that provides energy is an intermediate in the intestinal digestion of glycogen and starch and is found in germinating grains (and other plants and vegetables)
Catalog No. 50-311-2197 Supplier Selleck Chemical LLC Supplier No. S4496100MG
Maltose (Maltobiose Malt Sugar Beta-Maltose D-Maltose) monohydrate an non-essential nutrient that provides energy is an intermediate in the intestinal digestion of glycogen and starch and is found in germinating grains (and other plants and vegetables)