Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Research Products International Corp D-(+)-Maltose Monohydrate, 5 Kilograms
SDP

Supplier:  Research Products International Corp M220005000.0

Encompass_Preferred

CAS Number: 6363-53-7
Molecular Formula: C12H22O11H2O
Molecular Weight: 360.3

Catalog No. 50-213-442


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.